Learn More
FAM208A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA553556
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: GKWQWKVHCK FQKKLKELGR LNAKALSLLT LLNVYQKKHL VEILSYHNCD SQTRNAPELD CLIRLQAQNI QQRHIVFLTE KNIKMLSSYT DNGIVVATAE DFMQNFKNLV GYHNSITEEN LPQLGANENL ES Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 90%.
Spezifikation
FAM208A | |
Polyclonal | |
Unconjugated | |
FAM208A | |
FAM208A | |
Recombit protein corresponding to Human FAM208A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UK61 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23272 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Research Use Only