missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM227B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA543959
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: EYSLILQNHT SEIFKWKSMI SETSSYRKLE RYGEFLKKYH KKKKIMLSDE Sequence homology: Human: 100%
FAM227B Polyclonal antibody specifically detects FAM227B in Human samples. It is validated for Western Blot.Spezifikation
FAM227B | |
Polyclonal | |
Unconjugated | |
FAM227B | |
C15orf33; FAM227B; family with sequence similarity 227 member B; family with sequence similarity 227, member B; protein FAM227B | |
Rabbit | |
Affinity Chromatography | |
RUO | |
196951 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96M60 | |
FAM227B | |
Synthetic peptide directed towards the N-terminal of human FAM227B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |