Learn More
FANCI Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA559014
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: YGKGVLSGEE CKKQLINTLC SGRWDQQYVI QLTSMFKDVP LTAEEVEFVV EKALSMFSKM NLQEIPPLVY QLLVLSSKGS RKSVLEGIIA FFSALDKQHN EEQSGDELLD VVTVPSGE Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 73%.
The Fanconi anemia complementation group (FANC) currently includes FANCA, FANCB, FANCC, FANCD1 (also called BRCA2), FANCD2, FANCE, FANCF, FANCG, FANCI, FANCJ (also called BRIP1), FANCL, FANCM and FANCN (also called PALB2). The previously defined group FANCH is the same as FANCA. Fanconi anemia is a genetically heterogeneous recessive disorder characterized by cytogenetic instability, hypersensitivity to DNA crosslinking agents, increased chromosomal breakage, and defective DNA repair. The members of the Fanconi anemia complementation group do not share sequence similarity; they are related by their assembly into a common nuclear protein complex. This gene encodes the protein for complementation group I. Alternative splicing results in two transcript variants encoding different isoforms.Spezifikation
FANCI | |
Polyclonal | |
Unconjugated | |
FANCI | |
FANCI; Fanconi anemia complementation group I; Fanconi anemia group I protein; Fanconi anemia group I protein homolog; Fanconi anemia, complementation group I; KIAA1794; Protein FACI | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55215 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NVI1 | |
FANCI | |
Recombinant protein corresponding to Human KIAA1794 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only