Learn More
Invitrogen™ Faricimab Humanized Recombinant Human Monoclonal Antibody
Faricimab Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA560119
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Faricimab Recombinant Monoclonal Antibody for SPR, Flow, ELISA, Functional
Faricimab (RG7716) is a bispecific antibody pharmaceutical that simultaneously targets Angiopoietin-2 (Ang-2) and VEGF-A.
Spezifikation
| Faricimab Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| RG7716; RO6867461 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |