missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO18 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 14275-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Spezifikation
FBXO18 | |
Polyclonal | |
Unconjugated | |
FBXO18 | |
F box DNA helicase 1, F box only protein 18, F box protein, helicase, 18, FBH1, Fbx18, FBXO18 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
291293, 50755, 84893 | |
-20°C | |
Liquid |
Immunofluorescence, Western Blot, Immunocytochemistry | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8K2I9, Q8NFZ0 | |
Fbxo18 | |
FBXO18 Fusion Protein Ag5622 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |