missing translation for 'onlineSavingsMsg'
Learn More

FBXO7 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 10696-1-AP-20UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16897243

  • 140.00 € / 20 Mikroliter
Verfügbar ab: 15-05-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class and it may play a role in regulation of hematopoiesis. Alternatively spliced transcript variants of this gene have been identified with the full-length natures of only some variants being determined.
Spezifikation

Spezifikation

FBXO7
Polyclonal
Unconjugated
FBXO7
F box only protein 7, F box protein 7, FBX, FBX07, FBX7, FBXO7, PARK15, PKPS
Rabbit
Antigen Affinity Chromatography
RUO
25793, 69754
-20°C
Liquid
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q3U7U3, Q9Y3I1
Fbxo7
FBXO7 Fusion Protein Ag1035
20 μL
Primary
Human, Mouse
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote