Learn More
Instech Laboratories™ Feeding tube, mouse oral gavage, polyurethane, 16ga x 38mm, sterile
Artikelnummer. 15300350
Encompass
Ansicht ändern
Click to view available options
Produkttyp:
Zulaufschlauch
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Produkttyp
15300350 Zulaufschlauch
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 15300350 Lieferant Instech Laboratories™ Lieferanten-Nr. FTPU163850
Verfügbar ab: 09-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
These specialty tubes feature a larger inner diameter for easier delivery of viscous compounds and a straight tip that wipes clean to help in cases where aspiration of compounds would be hazardous to the animal. Availablein two sizes: 13ga for rats and 16ga for mice.
  • Plastic feeding tubes, 16ga x 38mm
  • For mouse oral gavage
  • Designed to minimise trauma to oesophagus
  • Polyurethane tube with rounded tip
  • Large internal diameter for viscous compounds
  • Straight tube wipes clean to prevent aspiration of compounds
  • Disposable
  • Supplied as 10 sterile pouches of 5 tubes
Spezifikation
Produkttyp Zulaufschlauch