Learn More
Instech Laboratories™ Feeding tube, rodent oral gavage, polypropylene, 20ga x 38mm, sterile
Artikelnummer. 15165024
Encompass
Ansicht ändern
Click to view available options
Produkttyp:
Zulaufschlauch
Packungsgröße:
50 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Produkttyp Packungsgröße
15165024 Zulaufschlauch 50 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 15165024 Lieferant Instech Laboratories™ Lieferanten-Nr. FTP203850
Verfügbar ab: 14-09-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Instech's polypropylene gavage tubes are flexible to reduce trauma, translucent to visualise delivery, and disposable to eliminate the cost of cleaning and the chance of compound crossover. Flexible tubes are a recommended animal welfare refinement over rigid metal tubes. The tube and luer hub are made of the same material used to make plastic syringes. The soft, bulb-shaped tip is designed to reduce the chances of inadvertent placement in the trachea and perforation of the oesophagus. Available in 11 sizes for rodents ranging from mouse pups to rats to rabbits.
  • Plastic feeding tubes, 20ga x 38mm
  • For rodent oral gavage
  • Designed to minimise trauma to oesophagus
  • Polypropylene female luer (yellow) and flexible polypropylene tube with soft elastomer tip
  • Disposable
  • Supplied as 10 sterile pouches of 5 tubes
Spezifikation
Produkttyp Zulaufschlauch