Learn More
Ibidi Female Luer Lock Connector
Artikelnummer. 16813963
Encompass_Preferred
Change view
Click to view available options
Female Luer Lock Connector
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer.
16813963 Female Luer Lock Connector
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 16813963 Lieferant Ibidi Lieferanten-Nr. 10825
Please to purchase this item. Need a web account? Register with us today!
Female Luer Lock Connector is suitable for male Luer or Luer Lock adapters.
  • Suitable for male Luer or Luer Lock adapters
  • natural Polypropylene
  • sterilized
Spezifikation
Material Polypropylen
Zur Verwendung mit Male Luer Adapter, Male Luer Lock Adapter
Verpackungsmenge 25 Pk.
Sterilität Steril
Produkttyp Female Luer Tubing Connector