missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FER Recombinant Human Protein
FER is a member of the FPS/FES family of non-receptor tyrosine kinases.
Marke: Invitrogen™ PV4076
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
FER regulates cell-cell adhesion and mediates signaling from the cell surface to the cytoskeleton via growth factor receptors.
Spezifikation
NP_005237 | |
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton™ X-100, 2mM DTT and 50% glycerol | |
1mg | |
RUO - research use only | |
FER, TYK3 | |
Human | |
His Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
60.3kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Catalytic | |
FER Kinase Family | |
Recombinant | |
Baculovirus |