missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ FGFR2 (Human) Recombinant Protein
Human FGFR2 full-length ORF ( AAH39243.2, 1 a.a. - 723 a.a.) recombinant protein with GST-tag at N-terminal.
Marke: Abnova™ H00002263-P01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
- Sequence: MGLTSTWRYGRGPGIGTVTMVSWGRFICLVVVTMATLSLARPSFSLVEDTTLEPEGAPYWTNTEKMEKRLHAVPAANTVKFRCPAGGNPMPTMRWLKNGKEFKQEHRIGGYKVRNQHWSLIMESVVPSDKGNYTCVVENEYGSINHTYHLDVVERSPHRPILQAGLPANASTVVGGDVEFVCKVYSDAQPHIQWIKHVEKNGSKYGPDGLPYLKVLKAAGVNTTDKEIEVLYIRNVTFEDAGEYTCLAGNSIGISFHSAWLTVLPAPGREKEITASPDYLEIAIYCIGVFLIACMVVTVILCRMKNTTKKPDFSSQPAVHKLTKRIPLRRQVSAESSSSMNSNTPLVRITTRLSSTADTPMLAGVSEYELPEDPKWEFPRDKLTLGKPLGEGCFGQVVMAEAVGIDKDKPKEAVTVAVKMLKDDATEKDLSDLASEMEMMKMIGKHENIINLLGACTQDGPLYVIVEYASKGNLREYLRARRPPGMEYSYDINRVPEEQMTFEDLVSCTYQLARGMEYLASQKCIHRDLAARNVLVTENNVMKIADFGLARDINNIDYYKKTTNGRLPVKWMAPEALFDRVYTHQSDVWSFGVLMWEIFTLGGSPYPGIPVEELLKLLKEGHRMDKPANCTNELYMMMRDCWHAVPSQRPTFKQLVEDLDRILTLTTNEEYLDLSQPLEQYSPSYPDTRSSCSSGDDSVFSPDPMPYEPCLPQYPHINGSVKT
Spezifikation
AAH39243.2 | |
fibroblast growth factor receptor 2 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FGFR2 | |
GST |
2263 | |
Wheat germ expression system | |
25 μg | |
BEK, BFR-1, CD332, CEK3, CFD1, ECT1, FLJ98662, JWS, K-SAM, KGFR, TK14, TK25 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |