Learn More
ACROBiosystems FITC-Labeled Human CD8 alpha&beta (CD8A&CD8B) Heterodimer Protein, His Tag&Tag Free
Marke:  ACROBiosystems CDA-HF2W4-200ug
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 17116191
  • 2661.24 € / 200 Mikrogramm
Verfügbar ab: 28-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Integral membrane glycoprotein that plays an essential role in the immune response and serves multiple functions in responses against both external and internal offenses. CD8A forms disulfide-linked heterodimers with CD8B at the cell surface. CD8A homodimer molecules also promote the survival and differentiation of activated lymphocytes into memory CD8 T-cells. Additionally, CD8B plays a critical role in thymic selection of CD8+ T-cells. A palmitoylation site in the cytoplasmic tail of CD8B chain contributes to partitioning of CD8 into the plasma membrane lipid rafts where signaling proteins are enriched.
FITC-Labeled Human CD8A&CD8B Heterodimer Protein, His Tag&Tag Free, produced by co-expression of CD8 alpha and CD8 beta, has a calculated MW of 19.5 kDa (CD8 alpha) and 22.1 kDa (CD8 beta). Subunit CD8 alpha is fused with a polyhistidine tag at the C-term
Spezifikation
Spezifikation
NP_001759.3 (CD8A)/NP_004922.1 (CD8B)
200 μg
Ser 22 - Asp 182 (CD8A) and Leu 22 - Pro 170 (CD8B)
CD8 alpha and beta,CD8A and CD8B
FITC
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE
19.5 kDa (CD8A) and 22.1 kDa (CD8B)
FITC-Labeled Human CD8A&CD8B; Heterodimer Protein, His Tag&Tag; Free (CDA-HF2W4) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Asp 182 (CD8A) and Leu 22 - Pro 170 (CD8B) (Accession # P01732-1 (CD8A) and P10966-1 (CD8B)). It is the FITC labeled form of Human CD8A&CD8B; Heterodimer Protein, His Tag&Tag; Free (CDA-H52W5).
-20°C, avoid light
CD8 alpha and beta
Human
PBS, pH7.4
HEK293
Fluorescent labeled protein