Learn More
ACROBiosystems FITC-Labeled Human EGF R Protein, His Tag
Marke: ACROBiosystems EGR-HF2H5-200ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The epidermal growth factor receptor (EGFR; ErbB-1; HER1 in humans) is the cell-surface receptor for members of the epidermal growth factor family (EGF-family) of extracellular protein ligands. The epidermal growth factor receptor is a member of the ErbB family of receptors, a subfamily of four closely related receptor tyrosine kinases: EGFR (ErbB-1), HER2/c-neu (ErbB-2), Her 3 (ErbB-3) and Her 4 (ErbB-4). Mutations affecting EGFR expression or activity could result in cancer.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 70.5 kDa. The protein migrates as 85-105 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
NP_005219.2 | |
200 μg | |
Leu 25 - Ser 645 | |
EGFR,ERBB,ERBB1,HER1,PIG61,mENA | |
FITC | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
70.5 kDa | |
FITC-Labeled Human EGF R, His Tag (EGR-HF2H5) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accession # P00533-1). It is the FITC labeled form of Human EGF R, His Tag (EGR-H5222). | |
-20°C, avoid light | |
EGF R | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Fluorescent labeled protein |