missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems FITC-Labeled Human Mesothelin / MSLN (296-580) Protein, His Tag
387.09 € - 2661.24 €
Spezifikation
Zugriffsnummer | AAH09272 |
---|---|
Molekulargewicht | 33.0 kDa |
Quelle | FITC-Labeled Human Mesothelin (296-580), His Tag (Cat. No. MSN-HF223) is expressed from human HEK293 cells. It contains AA Glu 296 - Gly 580 (Accession # AAH09272). It is the FITC labeled form of Human Mesothelin (296-580), His Tag (Cat. No. MSN-H5223). |
Immunogen | Glu 296 - Gly 580 |
Lagerungsbedingungen | -20°C, avoid light |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16487879
|
ACROBiosystems
MSN-HF223-25UG |
25 μg |
387.09 €
25 Mikrogramm |
Verfügbar ab: 28-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16477879
|
ACROBiosystems
MSN-HF223-200UG |
200 μg |
2661.24 €
200 Mikrogramm |
Verfügbar ab: 28-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Mesothelin (MSLN) is also known as CAK1 antigen, Pre-pro-megakaryocyte-potentiating factor, which belongs to the mesothelin family. Mesothelin / MSLN can be proteolytically cleaved into the following two chains by a furin-like convertase: Megakaryocyte-potentiating factor (MPF) and the cleaved form of mesothelin. Both MPF and the cleaved form of mesothelin are N-glycosylated. Mesothelin / MSLN can interacts with MUC16. The membrane-anchored forms of MSLN may play a role in cellular adhesion. MPF potentiates megakaryocyte colony formation in vitro.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 33.0 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH09272 | |
FITC-Labeled Human Mesothelin (296-580), His Tag (Cat. No. MSN-HF223) is expressed from human HEK293 cells. It contains AA Glu 296 - Gly 580 (Accession # AAH09272). It is the FITC labeled form of Human Mesothelin (296-580), His Tag (Cat. No. MSN-H5223). | |
-20°C, avoid light | |
Mesothelin | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Fluorescent labeled protein |
33.0 kDa | |
Glu 296 - Gly 580 | |
MSLN,Mesothelin,MPF | |
FITC | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |