Learn More
BUCHI FlashPure Empty Solid Loader: Parts
Artikelnummer. 16360316 Alle ansehen Buchi Produkte
Ansicht ändern
Click to view available options
FlashPure Empty Solid Loader Frits; Size: 12 g; Quantity: 20 pcs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer.
16360316 FlashPure Empty Solid Loader Frits; Size: 12 g; Quantity: 20 pcs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 16360316 Lieferant BUCHI Lieferanten-Nr. 148634551
Verfügbar ab: 03-11-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
Parts for FlashPure Empty Solid Loaders. FlashPure Solid Loaders are ideal for samples that are not readily soluble in the desired mobile phase when performing flash chromatography. The sample is adsorbed onto a solid phase (silica or Celite) and then added to the empty solid loader tube.
  • Screw cap makes sealing easy
  • Various sizes for differing sample amounts
  • Choose a size large enough for the sample
  • Insertion Rod allows frit placement and loading of samples smaller than the entire volume
TRUSTED_SUSTAINABILITY
Spezifikation
Material Polyethylen
Modell Frits
Typ Fritte
Enthält 20 Stk.
Größe (metrisch) Röhrchengestell 12 g
Säulentyp Empty
Zur Verwendung mit (Anwendung) Solid Loading of Insoluble Samples
Zur Verwendung mit (Geräte) Reveleris Solid Loading