Learn More
R Langenbrinck™ g-TUBE™ for DNA Shearing
Artikelnummer. 15240099
Encompass
Change view
Click to view available options
Menge:
10 Röhrchen, einzeln verpackt
Packungsgröße:
1 Stück
Artikelnummer. Menge unitSize
15240099 10 Röhrchen, einzeln verpackt 1 Stück
1 options
Artikelnummer. 15240099
Marke: R Langenbrinck™ 520079
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
A single-use device that shears genomic DNA into selected fragment sizes ranging from 6kb to 20kb. The only equipment needed is a compatible bench-top centrifuge.
  • Uses centrifugal force to push the sample through a precisely manufactured orifice, producing the shearing forces in the sample that fragment DNA
  • Fragment size is selected by adjusting the centrifuge rotor speed which alters the flow rate through the orifice and thus the shearing force
  • Higher centrifugation RPM will produce shorter fragments and the protocol provided by Covaris contains the settings to generate fragment distributions centered on 6, 8, 10 or 20kbp
  • The entire process takes only 3 minutes and up to 24 samples can be processed simultaneously with a high reproducibility
Spezifikation
Puffer DI Water or TE or 10 mM Tris-Cl pH 8.5
Menge 10 Röhrchen, einzeln verpackt
Trennungsbereich 6 kb to 20 kb