Learn More
Saint-Gobain GA Rubber Tubing
Marke: Saint-Gobain 70203010
Weitere Details : Netto-Gewicht : 0.80000kg
Spezifikation
Made according to GLP requirements, FDA compliant | |
Rubber | |
10 m | |
General laboratory use | |
Amber | |
70°C |
2 mm | |
4 mm | |
1 mm | |
-25°C | |
Flexible |