Learn More
Galectin 9 Monoclonal Antibody (RG9-35), PE-Cyanine7, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
173.00 € - 414.00 €
Spezifikation
Antigen | Galectin 9 |
---|---|
Klon | RG9-35 |
Konzentration | 0.2 mg/mL |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15913254
|
Affymetrix eBioscience
25-9211-80 |
25 μg |
173.00 €
25 Mikrogramm |
Verfügbar ab: 22-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15923254
|
Affymetrix eBioscience
25-9211-82 |
100 μg |
414.00 €
100 Mikrogramm |
Verfügbar ab: 14-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Description: This RG9-35 monoclonal antibody recognizes mouse Galectin 9, a member of the S-type lectin family. Galectin 9 is widely expressed within a majority of hematopoietic cells with the particularly high expression on CD45R(B220)-intermediate pre-B cells. Applications Reported: This RG9-35 antibody has been reported for use in flow cytometric analysis. Applications Tested: This RG9-35 antibody has been tested by flow cytometric analysis of mouse bone marrow cells using the Intracellular Fixation & Permeabilization Buffer Set (Product No. 88-8824) and protocol. Please refer to Best Protocols: Protocol A: Two step protocol for (cytoplasmic) intracellular proteins located under the Resources Tab online. This may be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product No. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product No.
Galectin 9 Monoclonal antibody specifically detects Galectin 9 in Mouse samples. It is validated for Flow Cytometry.Spezifikation
Galectin 9 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
O08573 | |
Lgals9 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Lgals9 |
RG9-35 | |
Flow Cytometry | |
PE-Cyanine7 | |
Rat | |
Mouse | |
16859 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |