missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GALNT9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 25967-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene encodes a member of the UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase (GalNAc-T) family of enzymes. GalNAc-Ts initiate mucin-type O-linked glycosylation in the Golgi apparatus by catalyzing the transfer of GalNAc to serine and threonine residues on target proteins. They are characterized by an N-terminal transmembrane domain, a stem region, a lumenal catalytic domain containing a GT1 motif and Gal/GalNAc transferase motif, and a C-terminal ricin/lectin-like domain. GalNAc-Ts have different, but overlapping, substrate specificities and patterns of expression. This gene is expressed specifically in the brain, with highest expression in the cerebellum. Multiple transcript variants encoding different isoforms have been found for this gene.Spezifikation
GALNT9 | |
Polyclonal | |
Unconjugated | |
GALNT9 | |
GALNAC T9, GALNT9, pp GaNTase 9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
231605, 304571, 50614 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9HCQ5 | |
GALNT9 | |
GALNT9 Fusion Protein Ag23233 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |