missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAPDH Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 33 publications
212.10 € - 509.25 €
Spezifikation
| Antigen | GAPDH |
|---|---|
| Anwendungen | Immunofluorescence, Immunohistochemistry, Western Blot, Immunocytochemistry |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Form | Antisera |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
18033316
|
Novus Biologicals
NB300-327-0.025ML |
0.025 mL |
225.00 € 212.10 € / 25 Mikroliter Sparen 12.90 € 5% Rabatt |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | |||||
|
18709753
|
Novus Biologicals
NB300-327 |
0.1 mL |
539.00 € 509.25 € / 0.10 Milliliter Sparen 29.75 € 5% Rabatt |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | |||||
Beschreibung
GAPDH Polyclonal specifically detects GAPDH in Human, Mouse, Rat, Porcine, Bacteria, Bovine, Chicken, Equine, Fungi, Invertebrate, Yeast samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Spezifikation
| GAPDH | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Rat, Chicken, Mouse, Pig, Yeast, Equine, Fungi, Bovine, Invertebrate, Bacteria, Human | |
| aging-associated gene 9 protein, EC 1.2.1, EC 1.2.1.12, EC 2.6.99.-, G3PD, GAPD, glyceraldehyde 3-phosphate dehydrogenase, glyceraldehyde-3-phosphate dehydrogenase, MGC88685, Peptidyl-cysteine S-nitrosylase GAPDH | |
| GAPDH | |
| IgG | |
| Unpurified | |
| Human, cow, pig, mouse, rat, bird and chicken. Yeast reactivity reported in scientific literature (PMID: 23709183). Since GAPDH is one of the most conserved proteins known and it is therefore likely that the antibody will react with other species also. Invertebrate reactivity reported in scientific literature (PMID: 18638509). |
| Immunofluorescence, Immunohistochemistry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Alzheimers Research, Apoptosis, Autophagy, Cancer, DNA Repair, Lipid and Metabolism, Loading Controls, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Nuclear Receptors Coactivators and Corepressors, Signal Transduction, Translation Control | |
| P10096 | |
| 2597 | |
| This GAPDH antibody was developed against full length recombinant human GAPDH | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 36 kDa |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts