Learn More
3M™ Gas-, Dampf- und Partikelfilter, Serie 6000
Provides excellent field of vision as a result of the unique trapezoidal shape. The 3M Gas, Vapor, and Particulate Filter uses bayonet-style cartridge connection for fitting to 3M 6000 and 7907 Series Full Face Masks.
Brand: 3M™ 1469SB
Additional Details : Netto-Gewicht : 0.99000kg
Description
Unique, compatible Bayonet Filter Attachment System allows different types of filters to just be clicked into place ensuring optimum ease of use.- Lightweight, low breathing resistance, and well-balanced when fitted to mask
- Provides excellent field of vision as a result of the unique trapezoidal shape
- Bayonet-style cartridge connection for fitting to 3M 6000 and 7907 Series Full Face Masks
Specifications
Single compound organic vapor (boiling point below 65°C) and particulates | |
AXP3 R | |
Typical industrial applications for filters are laboratories and particulate applications | |
2/Pk. |
EN14387:2004 + A1:2008, CE Approved | |
Gas, Vapor and Particulate Filter | |
3M™ 6000 and 3M™ 7907 Series Full Face Masks |