Learn More
GCLC Antibody (S08-2F5), Novus Biologicals™
Rabbit Monoclonal Antibody
Marke:  Novus Biologicals NBP3-19644-100ul
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 18646853
  • 645.00 € / 100 Mikroliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
GCLC Monoclonal antibody specifically detects GCLC in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
GCLC
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot
S08-2F5
Unconjugated
EC 6.3.2.2, gamma-ECS, Gamma-glutamylcysteine synthetase, GCS, GCS heavy chain, GLCL, GLCLCGCL, glutamate--cysteine ligase catalytic subunit, glutamate-cysteine ligase, catalytic subunit
A synthetic peptide of human GCLC (Uniprot # P48506)
100 μg
Cell Biology, Lipid and Metabolism
2729
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.