missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCLM Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
214.00 € - 584.00 €
Spezifikation
| Antigen | GCLM |
|---|---|
| Verdünnung | Western Blot 1:500 - 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
| Anwendungen | ELISA, Western Blot, Immunocytochemistry/Immunofluorescence |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Product Code | Brand | Menge | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Menge | Price | Quantity & Availability | |||||
|
30229577
|
Novus Biologicals
NBP3-38134-20ul |
20 μL |
214.00 €
20 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
30226920
|
Novus Biologicals
NBP3-38134-100ul |
100 μL |
584.00 €
100 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Description
GCLM Polyclonal antibody specifically detects GCLM in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ImmunofluorescenceSpecifications
| GCLM | |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| PBS (pH 7.3), 50% glycerol | |
| 2730 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Gamma-ECS regulatory subunit, Gamma-glutamylcysteine synthetase regulatory subunit, GCS light chain, GLCLRglutamate--cysteine ligase regulatory subunit, glutamate-cysteine ligase (gamma-glutamylcysteine synthetase), regulatory(30.8kD), Glutamate--cysteine ligase modifier subunit, glutamate-cysteine ligase regulatory protein, glutamate-cysteine ligase, modifier subunit, GSC light chain | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-274 of human GCLM (NP_002052.1).,, Sequence:, AQLDSVIIASPPIEDGVNLSLEHLQPYWEELENLVQSKKIVAIGTSDLDKTQLEQLYQWAQVKPNSNQVNLASCCVMPPDLTAFAKQFDIQLLTHNDPKELLSEASFQEALQESIPDIQAHEWVPLWLLRYSVIVKSRGIIKSKGYILQAKRRGS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title