missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDF11 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA550631
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total GDF11 protein.
GDF11 Polyclonal antibody specifically detects GDF11 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
GDF11 | |
Polyclonal | |
Unconjugated | |
GDF11 | |
BMP11; BMP-11; bone morphogenetic protein 11; GDF11; GDF-11; growth differentiation factor 11; growth/differentiation factor 11 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10220, 14561 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.7 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
O95390, Q9Z1W4 | |
GDF11 | |
Synthetic peptide corresponding to residues near the C terminal of human growth differentiation factor 11 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |