missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gelsolin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA519848
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
For Western Blot, this antibody has non-specific bands at 120 kDa,140 kDa and 52 kDa.
The protein encoded by this gene binds to the "e;plus"e; ends of actin monomers and filaments to prevent monomer exchange. The encoded calcium-regulated protein functions in both assembly and disassembly of actin filaments. Defects in this gene are a cause of familial amyloidosis Finnish type (FAF). Multiple transcript variants encoding several different isoforms have been found for this gene.Spezifikation
Gelsolin | |
Polyclonal | |
Unconjugated | |
GSN | |
Actin-depolymerizing factor; ADF; ADFBrevin; AGEL; Brevin; DKFZp313L0718; gelsolin; gelsolin (amyloidosis, Finnish type); Gsb; Gsn; hypothetical protein LOC535077; plasma gelsolin precursor; RP11-477J21.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
227753, 2934, 296654 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 1% BSA and 0.02% sodium azide; pH 7.4 | |
P06396, P13020, Q68FP1 | |
GSN | |
Synthetic peptide conjugated to KLH derived from within residues 750 to the C-terminus of Human Gelsolin. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |