Learn More
GFOD2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA559360
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: PVSMAASFED GLYMQSVVDA IKRSSRSGEW EAVEVLTEEP DTNQNLCEAL QRNNL Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 91%.
Promotes matrix assembly.Spezifikation
GFOD2 | |
Polyclonal | |
Unconjugated | |
GFOD2 | |
5730466C23Rik; GFOD2; glucose-fructose oxidoreductase domain containing 2; glucose-fructose oxidoreductase domain-containing protein 2; RGD1311198; UNQ9430/PRO34691 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
81577 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q3B7J2 | |
GFOD2 | |
Recombinant protein corresponding to Human GFOD2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only