Learn More
GFP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA1980A
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-980A detects Green Fluorescent Protein (GFP). PA1-980A has been successfully used in Western blot and immunofluoresence procedures. By Western blot, this antibody detects recombinant GFP. PA1-980A immunizing peptide corresponds to the N-terminal amino acid residues 26-39 of Aequorea victoria wild-type GFP. This sequence is completely conserved between GFP and the modified and enhanced Blue, Green, and Yellow FP variants. PA1-980A immunizing peptide (Cat. # PEP-033) is available for use in neutralization and control experiments.
GFP Polyclonal antibody specifically detects GFP in Tag samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
GFP | |
Polyclonal | |
Unconjugated | |
GFP; GFP tag; GFP2; green fluorescence; green fluorescent; Turbo eGFP | |
Synthetic Peptide: H(26) K F S V S G E G E G D A T(39) C | |
100 μL | |
Primary | |
Antibody | |
IgG |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Tag |