missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ebro Electronic™ GFX 460 B Kontaktthermometer
Kontaktthermometer, Pt100, ohne Fühler, Lemo Größe 1
Marke: Ebro Electronic™ 1340-5464A
Weitere Details : Netto-Gewicht : 0.20000kg
Beschreibung
Sonde PT100 für TFX 430,190mm, glasummanteltSpezifikation
EMC-Directive 2014/30/EU; EN 61326-1:2013; RoHS Directive 2011/65/EU; Electromagnetic Compatibility Regulations 2016 BS EN 61326-1:2013; The Restriction of the Use of Certain Hazardous Substances in Electrical and Electronic Equipment Regulations 2012 | |
Electronic Contact Thermometer, for replaceable Pt 100 probes | |
Thermometer, calibration certificate, instruction manual | |
GFX 460 B | |
1 °C | |
None | |
300 °C | |
-50 °C | |
Factory Calibrated | |
±1 °C | |
50 mm |
Single-use | |
52 mm | |
335 mm | |
DIN 45322 Diodenstecker | |
0.24 kg | |
572°F | |
-58°F | |
-50°bis +300 °C | |
TPX 130, TPX 230, TPX 330, AX 110, AG 151, AX 400 | |
Grün |