missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GGT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA561058
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: LLPNVTTVER NIDQAVTAAL ETRHHHTQI Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 41%.
Glutamyltransferase (gGT) from human kidney is a membrane-bound enzyme that transfers the gamma-glutamly moiety of gamma-glutamyl compounds such as glutathione to an acceptor, which may be an amino acid or a di- or tripeptide or water. The principal functions of gGT may be hydrolysis and metabolism of glutathione. Various human gGT isoforms have been found. The molecular mass of gGT for normal kidney is 90kDa and gGT can be used as a renal cell marker.Spezifikation
GGT1 | |
Polyclonal | |
Unconjugated | |
GGT1 | |
CD224; D22S672; D22S732; dwg; gamma glutamyl-transferase 1; gamma-glutamyl transpeptidase; gamma-glutamyltransferase 1; Gamma-glutamyltranspeptidase 1; GGLUT; Ggt; GGT 1; Ggt1; GGT-1; Ggtp; glutathione hydrolase 1; Glutathione hydrolase 1 heavy chain; Glutathione hydrolase 1 light chain; Glutathione hydrolase 1 proenzyme; GTG; Leukotriene-C4 hydrolase; testicular tissue protein Li 73 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2678 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P19440 | |
GGT1 | |
Recombinant protein corresponding to Human GGT1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only