Learn More
Ghrelin Polyclonal Antibody, Invitrogen™
Chicken Polyclonal Antibody
Marke: Invitrogen PA19596
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-9596 detects Ghrelin in human and mouse samples. PA1-9596 has been successfully used in ELISA procedures. PA1-9596 immunogen is a synthetic peptide corresponding to amino acids 17-28 of human ghrelin.
GHRL encodes ghrelin-obestatin preproprotein, which generates ghrelin and obestatin. Ghrelin is an the ligand for the growth hormone secretagogue receptor type 1 and is involved in regulating growth hormone release. Ghrelin has an appeptite-stimulating effect and induces adiposity and stimulates gastric acid secretion. Obestatin may be the ligand for the orphan G protein-coupled receptor GPR39 and is involved in satiety and can have an appetite-reducing effect resulting in decreased food intake.Spezifikation
Ghrelin | |
Polyclonal | |
Unconjugated | |
GHRL | |
2210006E23Rik; AMCBX1; Appetite-regulating hormone; appetite-regulating hormone; uncharacterized protein LOC58991; exon 2- and 3-deleted preproghrelin; exon 2-deleted preproghrelin; FIP3; FIP-3; Fip3p; Ghr; Ghrelin; ghrelin 27; ghrelin 28; ghrelin and obestatin prepropeptide; ghrelin precursor; ghrelin, growth hormone secretagogue receptor ligand; ghrelin/obestatin; ghrelin/obestatin preprohormone; ghrelin/obestatin prepropeptide; Ghrelin-27; Ghrelin-28; ghrelin-like protein; GHRL; Growth hormone secretagogue; growth hormone-releasing peptide; IKK-gamma; In2c-preproghrelin; IP; IP1; IP2; IPD2; m46; Motilin-related peptide; MTLRP; MTLRPAP; NEMO; Obestatin; Obestatin-13; Obestatin-23; OTTHUMP00000207794; OTTHUMP00000207795; OTTHUMP00000207796; OTTHUMP00000207798; prepro-appetite regulatory hormone; preproghrelin; proghrelin; Protein M46; UNQ524/PRO1066 | |
Chicken | |
Affinity chromatography | |
RUO | |
51738, 58991 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q9EQX0, Q9UBU3 | |
GHRL | |
Synthetic peptide corresponding to amino acids 17-28 of human ghrelin. | |
200 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgY |