Learn More
Huber Glass Accessory Set, Cold Trap
Beschreibung
- Glass accessory set for cold trap CT50 Single OLÉ includes:
- 503344 glass trap body
- 503345 three-way vacuum stopcock adaptor
- 503346 1-ltr receiver flask
- 503347 quick release clamp 100 mm
- 504544 O-Ring DN100
Spezifikation
Spezifikation
| Material | Glass |
| Typ | Accessory Kit |