Learn More
STEMCELL Technologies GloCell™ Fixable Viability Dye UV 450
Click to view available options
Menge:
100 Tests
5 x 100 Tests
Packungsgröße:
5 Stück
Beschreibung
- GloCell™ Fixable Viability Dye UV 450 is a fluorescent cell viability dye for staining live/dead mammalian cells in applications such as flow cytometry
- The dye irreversibly binds intracellular and cell surface amine groups and can be utilized prior to fixation, permeabilization, or cryopreservation
- Cells with compromised plasma membranes become permeable to the GloCell™ dye, resulting in a greater fluorescence compared to live cells in a cell viability assay
- GloCell™ Fixable Viability Dye UV 450 is excited by the UV laser at 355 nm and it has a fluorescence emission maximum of 450 nm that can be detected using the 450/50 band pass filter used to detect DAPI, Hoechst 33258, etc
Spezifikation
Spezifikation
| Zur Verwendung mit (Anwendung) | Flow Cytometry; Immunofluorescence |
| Menge | 100 Tests |