Learn More
STEMCELL Technologies GloCell™ Fixable Viability Dye Violet 450
Click to view available options
Menge:
100 Tests
5 x 100 Tests
missing translation for 'unitSize'
5 Stück
Description
- GloCell™ Fixable Viability Dye Violet 450 is a fluorescent cell viability dye for staining live/dead mammalian cells in applications such as flow cytometry
- The dye irreversibly binds intracellular and cell surface amine groups and can be utilized prior to fixation, permeabilization, or cryopreservation
- Cells with compromised plasma membranes become permeable to the GloCell™ dye, resulting in a greater fluorescence compared to live cells in a cell viability assay
- GloCell™ Fixable Viability Dye Violet 450 is excited by the violet laser at 405 nm and it has a fluorescence emission maximum of 450 nm that can be detected using the 440/40 or 440/50 band pass filter used to detect Pacific Blue™
Specifications
Specifications
| Zur Verwendung mit (Anwendung) | Flow Cytometry; Immunofluorescence |
| Menge | 100 Tests |