missing translation for 'onlineSavingsMsg'
Learn More
Learn More
glycerol-3-phosphate permease Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marke: Novus Biologicals NBP1-87524-25ul
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
glycerol-3-phosphate permease Polyclonal specifically detects glycerol-3-phosphate permease in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Spezifikation
| glycerol-3-phosphate permease | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| FLJ22340, G-3-P permease, G-3-P transporter, G3PPgene similar to glycerol-3-phosphate permease10Glycerol-3-phosphate permease, glycerol-3-phosphate transporter, solute carrier family 37 (glycerol-3-phosphate transporter), member 1, Solute carrier family 37 member 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 54020 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| SLC37A1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:IVKGELHKYCTAWDEADVRFSSQNRKSGSAAPHQLPDNETDCGWAPFDKNNY | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur