Learn More
Invitrogen™ GMNN Recombinant Rabbit Monoclonal Antibody (24GB4610)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA557030
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
GMNN Recombinant Monoclonal Antibody for Western Blot
GMNN inhibits DNA replication by preventing the incorporation of MCM complex into prereplication complex (pre-RC). It is degraded during the mitotic phase of the cell cycle. Its destruction at the metaphase-anaphase transition permits replication in the succeeding cell cycle.
Spezifikation
| GMNN | |
| Recombinant Monoclonal | |
| Unconjugated | |
| GMNN | |
| AW546347; Gem; geminin; Geminin GMNN; geminin, DNA replication inhibitor; GMNN; MGORS6; OTTHUMP00000039393; RP3 369A17.3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 291137, 51053 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 24GB4610 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75496 | |
| GMNN | |
| Recombinant protein of human Geminin (aa 110-209). | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |