Learn More
Goat anti-Human IgM Secondary Antibody, Invitrogen™
Goat Polyclonal Secondary Antibody
Marke: Invitrogen 31124
238.33 EUR gültig bis 2024-05-31
Benutzen Sie den Promotions-Code "21664" um Ihren Promotions-Preis zu erhalten.
Weitere Details : Netto-Gewicht : 0.80000kg
Beschreibung
Product # 31124 has been successfully used in Western blot, IF, ICC, IHC, ELISA, and ISH applications. The activity measured by a solid phase binding assay is 91%. Product # 31124 reacts with the μ chains of human IgM. Based on solid-phase enzyme immunoassay, this antibody cross-reacts with dog IgM (12%) and mouse IgM (12%). It has minimum cross-reactivity with immunoglobulins from other species. Reconstitute with 0.5 mL of distilled water (1.0 mg/mL after restoration).
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Spezifikation
Human IgM | |
Polyclonal | |
Unconjugated | |
Goat | |
500 μg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Glycerol (1:1) may be added for added stability. | |
IgG |
ELISA, Immunocytochemistry, Immunohistochemistry, In Situ Hybridization (ISH), Western Blot | |
1 mg/mL | |
PBS with 0.08% sodium azide; pH 7.8 | |
Affinity chromatography | |
RUO | |
Human | |
Lyophilized |