Learn More
Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, Biotin, Invitrogen™
Goat Polyclonal Secondary Antibody
Marke: Invitrogen 31802
Weitere Details : Netto-Gewicht : 0.00150kg
Beschreibung
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31802 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31802 reacts with the heavy chains of mouse IgG and with the light chains common to most mouse immunoglobulins, but does not react against non-immunoglobulin serum proteins. The antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reaction with human, bovine and horse serum proteins. However, these antibodies may cross-react with immunoglobulins from other species. Store product at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Reconstitute with 1.5 mL of distilled water (1.2 mg/mL after restoration). Country of Origin: USA
Mouse IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Mouse IgG (H+L) Cross-Adsorbed in Mouse samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot.Spezifikation
Mouse IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
Biotin | |
Goat | |
1.5 mL | |
Secondary | |
4° C | |
IgG |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
1.2 mg/mL | |
PBS with 15mg/mL BSA and 0.05% sodium azide; pH 7.6 | |
Affinity chromatography | |
RUO | |
Mouse | |
Lyophilized |