missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgG1 Cross-Adsorbed Secondary Antibody, Invitrogen™
Goat Polyclonal Secondary Antibody
Marke: Invitrogen A10538
138.70 EUR gültig bis 2024-05-31
Benutzen Sie den Promotions-Code "21664" um Ihren Promotions-Preis zu erhalten.
Weitere Details : Netto-Gewicht : 0.00450kg
Beschreibung
Mouse IgG1 Cross-Adsorbed Polyclonal antibody specifically detects Mouse IgG1 Cross-Adsorbed in Mouse samples. It is validated for Immunocytochemistry.Spezifikation
Mouse IgG1 Cross-Adsorbed | |
Polyclonal | |
Unconjugated | |
Goat | |
purified | |
RUO | |
Mouse | |
Liquid |
Immunocytochemistry | |
2 mg/mL | |
PBS with 5mM sodium azide; pH 7.5 | |
IgG gamma 1 | |
500 μg | |
Secondary | |
4° C | |
IgG |