Learn More
Goat anti-Mouse IgG2a Cross-Adsorbed Secondary Antibody, FITC, Invitrogen™
Goat Polyclonal Secondary Antibody
Marke: Invitrogen 31634
Weitere Details : Netto-Gewicht : 0.80000kg
Beschreibung
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31634 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31634 reacts with the heavy chains of mouse IgG2a, but not with heavy chains of other mouse IgG subclasses or with light chains of most mouse immunoglobulins. This antibody does not react against mouse IgM, or against non-immunoglobulin serum proteins. The antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reaction with human, bovine, and rabbit serum proteins. However, these antibodies may cross-react with immunoglobulins from other species. Store product protected from light at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Fluorescein Amax= 492 nm; Emax= 520 nm. Fluorophore/Protein: 7.6 μg/mg; ∽ 3 moles FITC per mole IgG (lot-dependent). Reconstitute with 0.35 mL of distilled water. Country of Origin: USA
Mouse IgG2a Cross-Adsorbed Polyclonal antibody specifically detects Mouse IgG2a Cross-Adsorbed in Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry, Immunoprecipitation.Spezifikation
Mouse IgG2a Cross-Adsorbed | |
Polyclonal | |
FITC | |
Goat | |
500 μg | |
Secondary | |
4° C, store in dark | |
IgG |
Immunocytochemistry, Immunohistochemistry, Immunoprecipitation | |
1.6 mg/mL | |
PBS with 15mg/mL BSA and 0.05% sodium azide; pH 7.6 | |
Antigen affinity chromatography | |
RUO | |
Mouse | |
Lyophilized |