missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgG2a Secondary Antibody, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen PA128885
230.85 EUR gültig bis 2024-05-31
Benutzen Sie den Promotions-Code "21664" um Ihren Promotions-Preis zu erhalten.
Additional Details : Netto-Gewicht : 0.01000kg
Description
PA1-28885 detects Mouse IgG2a from mouse samples.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Mouse IgG2a | |
Polyclonal | |
Unconjugated | |
Goat | |
Antigen affinity chromatography | |
RUO | |
Mouse | |
Liquid |
ELISA, Flow Cytometry, Immunocytochemistry, Immunodiffusion, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Goats were immunized with highly purified mouse IgG2a in Freund’s adjuvant | |
500 μg | |
Secondary | |
4° C | |
IgG |