Learn More
Goat anti-Rat IgG (H+L) Secondary Antibody, WesternDot™ 625, Invitrogen™
Goat Polyclonal Secondary Antibody
Marke: Invitrogen W10818
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
WesternDot fluorescently labeled secondary antibodies deliver superior signal-to-noise ratios and exhibit comparable sensitivity to enhanced chemiluminescence (ECL)-based methods in western blot applications. No stripping and re-probing or multiple blots are required for multiple protein detection as commonly needed with ECL or colorimetric detection. Differently labeled WesternDot antibodies can be applied to a single blot for simultaneous detection of multiple proteins using standard gel or blot imaging platforms (a list of instruments that are compatible with WesternDot detection can be found in the user manual below). Our WesternDot antibodies utilize Qdot nanocrystals enhanced with VIVID technology, making them brighter than the original Qdot reagents. Additionally, they have reduced intensity differences between the different colors, simplifying multiplex applications.
Rat IgG (H+L) Polyclonal antibody specifically detects Rat IgG (H+L) in Rat samples. It is validated for Western Blot.Spezifikation
Rat IgG (H+L) | |
Polyclonal | |
50mM borate with 1M betaine and 0.05% sodium azide; pH 8.3 | |
Rat Immunoglobin | |
25 blots | |
Secondary | |
4° C, do not freeze | |
IgG |
Western Blot | |
WesternDot 625 | |
Goat | |
purified | |
RUO | |
Rat | |
Liquid |