missing translation for 'onlineSavingsMsg'
Learn More

Gold(I) chloride, 99.99%, BLD Pharm™

Product Code. 31031562
Encompass_Preferred
Change view
Click to view available options
Menge:
250 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge
31031562 250 mg
31015576 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 31031562 Supplier BLD Pharm Supplier No. BD132343250mg
 Eingeschränkter Artikel

Dieser Artikel erfordert eine Preisaktualisierung, bevor er bestellt werden kann.

Fordern Sie hier einen Preis für diesen Artikel an.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Metalsbasis

Specifications

Chemischer Name oder Material Gold(I) chloride
CAS 10294-29-8
Menge 250 mg
Summenformel AuCl
MDL-Nummer MFCD00046175
SMILES [Au+].[Cl-]
Molekulargewicht (g/mol) 232.42
Reinheit (%) 99.99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.