Learn More
GPR3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20847-1-AP-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The G-protein-coupled receptor (GPCR) superfamily is comprised of an estimated 600-1, 000 members and is the largest known class of molecular targets with proven therapeutic value. GPR3 is an orphan receptor with constitutive G signaling activity. GPR3 modulates a number of brain functions, including behavioral responses to stress, neurite outgrowth and amyloid-beta peptide generation suggesting that it may play a role in Alzheimer′s disease.Spezifikation
| GPR3 | |
| Polyclonal | |
| Unconjugated | |
| GPR3 | |
| ACCA, ACCA orphan receptor, G protein coupled receptor 3, GPR3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2827 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P46089 | |
| GPR3 | |
| GPR3 Fusion Protein Ag14257 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |