missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HACE1 Monoclonal Antibody (OTI4E10), TrueMAB™, OriGene
Description
HACE1 Monoclonal antibody specifically detects HACE1 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | HACE1 |
| Anwendungen | Western Blot |
| Klassifikation | Monoclonal |
| Klon | OTI4E10 |
| Konzentration | 1 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gen | CLEC4M |
| Gen-Zugriffsnummer | Q9H2X3 |
| Gen-Alias | 1700042J16Rik; A730034A22Rik; BC025474; E3 ubiquitin-protein ligase HACE1; hace 1; Hace1; HECT domain and ankyrin repeat containing E3 ubiquitin protein ligase 1; HECT domain and ankyrin repeat containing, E3 ubiquitin protein ligase 1; HECT domain and ankyrin repeat-containing E3 ubiquitin-protein ligase 1; HECT-type E3 ubiquitin transferase HACE1; KIAA1320; SPPRS |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?