Learn More
HCLS1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA540453
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: VNDISEKEQR WGAKTIEGSG RTEHINIHQL RNKVSEEHDV LRKKEMESGP Sequence homology: Cow: 86%; Dog: 86%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 86%; Rabbit: 86%; Rat: 86%
HCLS1 Polyclonal antibody specifically detects HCLS1 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
HCLS1 | |
Polyclonal | |
Unconjugated | |
HCLS1 | |
AW213261; CTTNL; HCLS1; hematopoietic cell specific Lyn substrate 1; hematopoietic cell-specific Lyn substrate 1; Hematopoietic lineage cell-specific protein; HS1; LckBP1; p75 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
15163, 3059 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P14317, P49710 | |
HCLS1 | |
Synthetic peptide directed towards the N-terminal of human HCLS1 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |