missing translation for 'onlineSavingsMsg'
Learn More

HDAC-IN-3, MedChemExpress

Product Code. 16491859
Encompass_Preferred
Change view
Click to view available options
Menge:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
1 Milligramm
50 Milligramm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge Unit Size
16491859 1 mg 1 Milligramm
16487898 10 mg -
16453847 50 mg 50 Milligramm
16427716 100 mg -
16420368 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16491859 Supplier Medchem Express Supplier No. HY19772
 Eingeschränkter Artikel

Dieser Artikel erfordert eine Preisaktualisierung, bevor er bestellt werden kann.

Fordern Sie hier einen Preis für diesen Artikel an.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress HDAC-IN-3 is a histone deacetylase (HDAC) inhibitor, extracted from patent WO/2008040934 A1.

HDAC-IN-3 is a histone deacetylase (HDAC) inhibitor, extracted from patent WO/2008040934 A1.

Specifications

Chemischer Name oder Material HDAC-IN-3
Physikalische Form Solid
CAS 1018673-42-1
Farbe Earth Yellow
Menge 1 mg
Hinweise zur Reinheitsqualität Research
Summenformel C22H33N3O4
Zur Verwendung mit (Anwendung) Cancer-programmed cell death
Löslichkeitsinformationen DMSO : 50 mg/mL (123.91 mM; Need ultrasonic)
SMILES O=C([C@@H](NCC1=CN=C(CCC(NO)=O)C=C1)C2CCCCC2)OC3CCCC3
Molekulargewicht (g/mol) 403.52
Formelmasse 403.52
Reinheit (%) 99.41%
Güte Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.