missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HE4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 14406-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene encodes a protein that is a member of the WFDC domain family. The WFDC domain, or WAP Signature motif, contains eight cysteines forming four disulfide bonds at the core of the protein, and functions as a protease inhibitor in many family members. This gene is expressed in pulmonary epithelial cells, and was also found to be expressed in some ovarian cancers. The encoded protein is a small secretory protein, which may be involved in sperm maturation.Spezifikation
HE4 | |
Polyclonal | |
Unconjugated | |
WFDC2 | |
HE4, WAP5, WFDC2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10406 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q14508 | |
WFDC2 | |
HE4 Fusion Protein Ag5684 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |