Learn More
HGD Monoclonal Antibody (1F1), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA524579
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Peptide Sequence: MAELKYISGF GNECSSEDPR CPGSLPEGQN NPQVCPYNLY AEQLSGSAFT CPRSTNKRSW LYRILPSVSH KPFESIDEGH VTHNWDEVDP DPNQLRWKPF EIPKASQKKV DFVSGLHTLC GAGDIKSNNG LAIHIFLCNT SMENRCFYNS DGDFLIVPQK GNLLIYTEFG KMLVQPNEIC VIQRGMRFSI DVFEETRGYI LEVYGVHLEL PDLGPIGANG LANPRDFLIP IAWYEDRQVP GGYTVINKYQ GKLFAAKQDV SPFNVVAWHG NYTPYKYNLK NFMVINSVAF DHADPSIFTV LTALRRPARS SWHLRGLPMA PWHLCLNHL
HGD Monoclonal antibody specifically detects HGD in Human samples. It is validated for ELISA, Immunoprecipitation, Western Blot.Spezifikation
HGD | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q93099 | |
HGD | |
HGD (AAH20792, 1 a.a. ∽ 329 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Immunoprecipitation, Western Blot | |
1F1 | |
Unconjugated | |
HGD | |
aku; alkaptonuria; HGD; HGO; homogentisate 1, 2-dioxygenase; homogentisate 1,2-dioxygenase; homogentisate oxidase; homogentisate oxygenase; Homogentisic acid oxidase; homogentisicase | |
Mouse | |
Protein A | |
RUO | |
3081 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |