Learn More
HGF Activator Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA563545
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: DERPWCYVVK DSALSWEYCR LEACESLTRV QLSPDLLATL PEPASPGRQA CGRRHKKRTF LRPRIIGGSS SLPGSHPWLA AIYIG Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 83%.
This gene encodes a member of the peptidase S1 protein family. The encoded protein is first synthesized as an inactive single-chain precursor before being activated to a heterodimeric form by endoproteolytic processing. It acts as serine protease that converts hepatocyte growth factor to the active form. Alternative splicing results in multiple transcript variants.Spezifikation
HGF Activator | |
Polyclonal | |
Unconjugated | |
Hgfac | |
hepatocyte growth factor activator; Hepatocyte growth factor activator long chain; Hepatocyte growth factor activator short chain; HGF activator; HGFA; Hgfac | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3083 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q04756 | |
Hgfac | |
Recombinant protein corresponding to Human HGF Activator | |
100 μL | |
Primary | |
Human | |
Antibody | |