missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIP1 Related Antibody (1E1), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 4 publications
199.00 € - 450.00 €
Spezifikation
| Antigen | HIP1 Related |
|---|---|
| Klon | 10 |
| Verdünnung | Western Blot 1:1000, Immunohistochemistry reported in scientific literature, Immunoprecipitation reported in scientific literature, Immunohistochemistry-Paraffin |
| Anwendungen | Western Blot, Immunohistochemistry, Immunoprecipitation |
| Klassifikation | Monoclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
18109219
|
Novus Biologicals
NB300-206SS |
0.025 mL |
199.00 €
25 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
18234060
|
Novus Biologicals
NB300-206 |
0.1 mL |
450.00 €
0.10 Milliliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
HIP1 Related Monoclonal specifically detects HIP1 Related in Human, Mouse, Rat, Bacteria samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Spezifikation
| HIP1 Related | |
| Western Blot 1:1000, Immunohistochemistry reported in scientific literature, Immunoprecipitation reported in scientific literature, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human, Mouse, Rat, Bacteria | |
| O75146 | |
| 9026 | |
| Human HIP1 Related-TALIN Homology Region (57 kDa). [UniProt# O75146] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| 10 | |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Biology, Neurodegeneration, Neuronal Cell Markers, Neuroscience | |
| Ascites with 0.1% Sodium Azide | |
| FLJ14000, HIP-12, HIP12HIP1-related protein, HIP3, huntingtin interacting protein 1 related, huntingtin interacting protein 12, Huntingtin-interacting protein 12, huntingtin-interacting protein 1-related protein, ILWEQ, KIAA0655FLJ27022, MGC47513 | |
| HIP1R | |
| IgG1 κ | |
| Unpurified | |
| 120 kDa |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts